Anti-IL6 Rabbit Polyclonal Antibody

GTX29626
89367-644EA 637.26 CAD
89367-644
Anti-IL6 Rabbit Polyclonal Antibody
Interleukin-6 is a potent lymphoid cell growth factor that affects B-lymphocytes, T lymphocytes, and hybridoma cells. It will also affect cytotoxic T cells in combination with other factors such as IL-2 and gamma-interferon.

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR