Anti-IL8 Chicken Polyclonal Antibody

GTX82044
89368-042EA 643.47 CAD
89368-042
Anti-IL8 Chicken Polyclonal Antibody
IL-8 is a chemotactic factor that attracts neutrophils. basophils. and T-cells. but not monocytes. It is also involved in neutrophil activation. It is released from several cell types in response to an inflammatory stimulus. IL-8(6-77) has a 5-10-fold higher activity on neutrophil activation. IL-8(5-77) has increased activity on neutrophil activation and IL-8(7-77) has a higher affinity to receptors CXCR1 and CXCR2 as compared to IL-8(1-77). respectively.

Type: Primary
Antigen: IL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR