Anti-INOS Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX73126
89353-146EA 565.28 CAD
89353-146
Anti-INOS Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to iNOS

Recommended Starting Dilutions: For IHC-P: Use at a dilution of 1:2,000 Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: INOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR