Anti-INOS Rabbit Polyclonal Antibody

GTX31049
89363-024EA 492.47 CAD
89363-024
Anti-INOS Rabbit Polyclonal Antibody
Produces nitric oxide (NO) which is a messenger molecule with diverse functions throughout the body. In macrophages, NO mediates tumoricidal and bactericidal actions.

Type: Primary
Antigen: INOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR