Anti-JNK2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13871
89359-880EA 742.11 CAD
89359-880
Anti-JNK2 Rabbit Polyclonal Antibody
Antibodies
c-Jun amino terminal kinases (JNKs) are MAP kinase family members that differ from other MAP kinases in that they are phosphorylated at the Thr-Pro-Tyr phosphorylation motif instead of the characteristic MAP kinase Thr-Glu-Tyr motif. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.

WB: Use at a concentration of 1 - 3 ?g/ml. Predicted molecular weight: 51 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: JNK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR