To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein A9
- Clonality:Monoclonal
- Clone:MAC387
- Gene ID:6280
- Host:Mouse
- Isotype:IgG1
- Reactivity:Baboon
- Antigen symbol:S100A9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Antigen synonyms:CAGB,MRP14,CFAG,NIF,MAC387,MIF,L1AG,60B8AG,CGLB,P14,LIAG
- Storage buffer:Buffer contains 0.09% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Human monocytes
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein G affinity purified
- Cat. no.:89366-140
Specifications
About this item
Mouse Monoclonal to Human Macrophages Sample
For Flow Cytometry: Use at a dilution of 1:50 - 1:100. For IHC-Fr and IHC-P: Use at a dilution of 1:100 - 1:200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.** This product requires protein digestion pre-treatment e.g. trypsin, 0.1% for 10 minutes or antigen retrieval using heat treatment prior to staining of paraffin sections.
Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: MAC387
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Monkey, Pig, Rat, Bb, Cat, Guinea Pig, Horse, Rabbit, Marmoset