Anti-LAMP2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX30881
89352-740EA 624.4 CAD
89352-740
Anti-LAMP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to LAMP2

For ICC: Use at a concentration of 10 µg/ml. For WB: Use at a concentration of 1-2 µg/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR