Anti-MDM2 Rabbit Polyclonal Antibody

GTX15471
89351-116EA 518.15 CAD
89351-116
Anti-MDM2 Rabbit Polyclonal Antibody
Human homolog of murine double minute-2 (MDM2) and p53 function in mutual regulation: activation of p53 results in expression of MDM2 which consequently inhibits the ability of wild type p53 to stimulate expression of target genes. MDM2 is frequently overexpressed in the nuclei of soft tissue sarcomas, and to a much lesser extent in a wide variety of malignancies of bladder, breast, testes, cervix, glial cells.

Type: Primary
Antigen: MDM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR