Anti-Melan A Rabbit Polyclonal Antibody

GTX15468
89351-110EA 538.97 CAD
89351-110
Anti-Melan A Rabbit Polyclonal Antibody
MART1 (Melanoma Antigen Recognized by T cells) or MelanA is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE1, MAGE3, tyrosinase, gp100, gp75, BAGE1, and GAGE1. Subcellular fractionation shows that MART1 is present in melanosomes and endoplasmic reticulum.

Type: Primary
Antigen: Melan A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR