Anti-MMP2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25704
89352-136EA 774.12 CAD
89352-136
Anti-MMP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MMP2

For ELISA: Use at a concentration of 0.1 - 1.0?g/ml. For IP: Use at a concentration of 3.0 - 5.0 ?g/extract from 10^6 cells. For WB: Use at a concentration of 0.5 - 2?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR