Goat Anti-Mouse IgG Polyclonal Antibody

GTX77307
89353-618EA 244.11 CAD
89353-618
Goat Anti-Mouse IgG Polyclonal Antibody
The affinity purified antibody was tested at 1 mg/ml against 1 ug/ml solutions of IgG, IgM, IgG and IgE. It reacts with all immunoglobulin classes by immunodiffusion and IEP techniques. This does not imply and equal balance of reactivity to all classes. F/P ratio: 3-6.Optimal conjugate working dilutions should be determined for your particular assay conditions. Goats were immunized with highly purified mouse IgGFAB isolated from normal mouse IgG. The resulting antiserum was immunoabsorbed against mouse Fc and the anti FAB immunoaffinity isolated from an immunoabsorbant column. The antibody was then conjugated with fluoresceine isothiocyanate (FITC)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR