Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex

GTX77302
89353-612EA 291.6 CAD
89353-612
Anti-IgG Goat Polyclonal Antibody
Antibodies
The affinity purified anti lambda reacts only with lambda containing immunoglobulins and Fab fragments by IEP and gel diffusion techniques. Goats were immunized with purified mouse lambda chains in Freund's adjuvant. The resulting antiserum was solid phase absorbed with a mouse IgG kappa-containing affinity support. The monospecific anti lambda antibodies were then isolated off an immunoaffinity column of Fab lambda

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR