Anti-Nefh Chicken Polyclonal Antibody

Supplier: GeneTex
GTX30667
89362-950EA 1291.79 CAD
89362-950
Anti-Nefh Chicken Polyclonal Antibody
Antibodies
Chicken Polyclonal antibody to NF-M

GTX30667 can be used in Western blot and immunofluorescence experiments. On blots GTX30667 runs ~145-170kDa. Suggested working dilutions: *Western Blot - 1:10,000 or lower (ECL) Immunofluorescence - 1:2,000*The investigator should determine the optimalworking dilution for a specific application.

Type: Primary
Antigen: Nefh
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR