Anti-NES Chicken Polyclonal Antibody

Supplier: GeneTex

GTX85457
89353-932EA 565.64 CAD
89353-932
Anti-NES Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to Nestin chicken (Neural stem cell marker)

Recommended starting dilutions: <br />For IHC: Use at 1:3000 - 1:6000.<br />For ICC/IF:Use at 1:3000 - 1:6000.<br />For WB: Use at 1:5000 - 1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR