Anti-RELA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74164
89365-398EA 944.11 CAD
89365-398
Anti-RELA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NFkB p65

Recommended Starting Dilutions: ELISA: 1/5000 - 1/25000 Gel Super Shift Assays: 0.5 - 1.0 ul per test Western Blotting: 1/2000 - 1/5000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR