Anti-NPYY2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX80745
89366-678EA 696.06 CAD
89366-678
Anti-NPYY2 Rabbit Polyclonal Antibody
Antibodies
This NPY Y2 antibody has been extensively characterized at the spinal level and is currently being characerized in brain.? The antibody has been tested in both rat and mouse and appears to work better on mose tissue, although positive staining is seen in rat.? For full characterizarion in dorsal root ganglion and spinal cord see: J Comp Neurol. 2005 Aug 29;489(3):328-48.

Type: Primary
Antigen: NPYY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR