Anti-ENO2 Mouse Monoclonal Antibody [clone: BBS/NC/VI-H14]

GTX75331
89365-752EA 304.2 CAD
89365-752
Anti-ENO2 Mouse Monoclonal Antibody [clone: BBS/NC/VI-H14]
Gamma enolase, is an enzyme involved in carbohydrate metabolism. It is found in mature neurons and cells of neuronal origin.

Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: BBS/NC/VI-H14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR