Anti-MAPK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX19329
89367-504EA 931.02 CAD
89367-504
Anti-MAPK Rabbit Polyclonal Antibody
Antibodies
p38 is a 38 kDa Stress Activated Protein Kinase / Map Kinase (SAPK / MAPK) that is fully activated by dual phosphorylation on threonine 180 and tyrosine 182, within the activation loop. p38 MAPK plays a critical role in the initiation of G2 delay after ultraviolet radiation; gene knock-out studies have also revealed a critical role for p38 in cardiac remodeling. Downstream targets of p38 include the transcription factors ELK1 and ATF2 and the kinases MAPKAPK2 and MAPKAPK5. p38 MAPK plays a role in the production of IL-6 and is thought to stabilize erythropoietin production during hypoxic stress. It is activated by environmental stress, many pro-inflammatory cytokines and lipopolysaccharide. Dual phosphorylation by MAP2K3 and MAP2K6 is required for activation of p38 MAPK. It interacts with MAX, Cdc25B, Cdc25C and binds to the kinase interaction domain in the protein tyrosine phosphatase PTPRR; this interaction retains p38 MAPK in the cytoplasm.

For IP: Use at a concentration of 10 µg/ml. For WB: Use at a concentration of 2µg/ml. Predicted molecular weight: 43 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR