Anti-P57KIP2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77602
89366-196EA 598.23 CAD
89366-196
Anti-P57KIP2 Rabbit Polyclonal Antibody
Antibodies
Kip2 p57 is a potent tight-binding inhibitor of several G1 cyclin/cdk complexes (cyclin E-cdk2, cyclin D2-cdk4, and cyclin A-cdk2) and, to a lesser extent, of the mitotic cyclin B-cdc2. It is a negative regulator of cell proliferation and may play a role in maintenance of the nonproliferative state throughout life.

Type: Primary
Antigen: P57KIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR