Anti-PDGFRB Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX25511
89361-662EA 765.37 CAD
89361-662
Anti-PDGFRB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PDGF Receptor beta

For ELISA: Use at a dilution of 1:900 - 1:1,100. For IHC-F and IHC-P: Use at a dilution of 1:50 - 1:100. For WB: Use at a dilution of 1:100 - 1:500. Detects a band of approximately 122 kDa (predicted molecular weight: 132 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PDGFRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR