Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX17039
89367-042EA 971.61 CAD
89367-042
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Protein gene product 9.5 (PGP9.5) is a member of a gene family whose products hydrolyze small C-terminal adducts of ubiquitin to generate the ubiquitin monomer. Expression of PGP9.5 is highly specific to neurons and to cells of the diffuse neuroendocrine system and their tumors. It is present in all neurons.

IF: Use at an assay dependent dilution. IHC-P: 1/25 - 1/50. Retrieve antigens with 70% Formic acid for 10-30 minutes at room temperature before commencing with IHC staining protocol. We suggest an incubation period of 30 minutes at room temperature. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR