Anti-PSEN1 Rabbit Polyclonal Antibody

GTX10281
89349-738EA 746.94 CAD
89349-738
Anti-PSEN1 Rabbit Polyclonal Antibody
Rabbit polyclonal to Presenilin 1

IP: Use at a dilution of 1/1000. WB: Use at a dilution of 1/1000 - 1/5000. Predicted molecular weight: 53 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR