Anti-PSEN1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX11146
89356-738EA 640.26 CAD
89356-738
Anti-PSEN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Presenilin 1

Recommended Starting Dilutions: Western Blotting: 1/500 - 1/1000. Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: PSEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR