Anti-PGR Mouse Monoclonal Antibody [clone: 4D8]

Supplier: GeneTex

GTX70226
89364-786EA 623.96 CAD
89364-786
Anti-PGR Mouse Monoclonal Antibody [clone: 4D8]
Antibodies
Mouse monoclonal [4D8] to Progesterone receptor

For WB: Use at a concentration of 1-5 µg/ml. Predicted molecular weight: 99 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR