Anti-S TAG Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77419
89353-698EA 624.13 CAD
89353-698
Anti-S TAG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S TAG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR