Anti-RPS6KB1 Rabbit Polyclonal Antibody

GTX22571
89361-292EA 466.68 CAD
89361-292
Anti-RPS6KB1 Rabbit Polyclonal Antibody
p70 S6 Kinase is a mitogen activated Ser/Thr protein kinase that is required for cell growth and G1 cell cycle progression. P70 S6 Kinase phosphorylates the S6 protein of the 40S ribosomal subunit and is involved in translational control of 5'oligopyrimidine tract mRNAs. A second isoform, p85 S6 Kinase, is derived from the same gene and is identical to p70 S6 Kinase except for 23 extra residues at the N-terminus that encode a nuclear localizing signal.

Type: Primary
Antigen: RPS6KB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR