Anti-SCF Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27316
89362-540EA 742.11 CAD
89362-540
Anti-SCF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SCF

ELISA: Use at a concentration of 0.5 ?g/ml. Allows detection of 0.2 - 0.4 ng/well of recombinant hSCF. Neut: Use at a concentration of 0.008 - 0.012 ?g/ml for [ND50] of the biological activity of hSCF (10 ng/ml). WB: Use at a concentration of 0.1 - 0.2 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR