Anti-SP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX26082
89361-818EA 910.59 CAD
89361-818
Anti-SP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SP1

WB: Use at a dilution of 1/200. Predicted molecular weight: 80 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR