Anti-T7 TAG Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX18611
89360-478EA 742.11 CAD
89360-478
Anti-T7 TAG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: T7 TAG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR