Anti-THRA Rabbit Polyclonal Antibody

Supplier: Genetex

GTX25621
89367-606EA 806.34 CAD
89367-606
Anti-THRA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Thyroid Hormone Receptor alpha 1

Recommended Starting Dilutions:IHC-P: 1:100 - 1:1000ICC/IF: 1:10 - 1:200ELISA: 1:2,000IHC: 1:100 - 1:400WB: 1:200 - 1:2500GSA: 1:10Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: THRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Chicken, Rat, Fish, Primates, Sheep, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR