Anti-TSH Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX38859
89363-058EA 1061.93 CAD
89363-058
Anti-TSH Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR