Anti-14-3-3 γ Mouse Monoclonal Antibody [clone: 3F8]

GTX14118
89359-946EA 507.03 CAD
89359-946
Anti-14-3-3 γ Mouse Monoclonal Antibody [clone: 3F8]
Members of the 14-3-3 family of proteins are highly conserved proteins, localized in neurons, and are axonally transported to the nerve terminals. They are also present, at lower levels, in various other eukaryotic tissues. 14-3-3 proteins appear to play important roles in a variety of signal transduction pathways, including those involved in cell cycle regulation and cell survival. Because 14-3-3 proteins bind to specific phosphoserine containing sequences they are likely to have an important role in signaling pathways mediated by serine/threonine protein kinases. 14-3-3 proteins regulate many cellular processes that are important in cancer biology, such as apoptosis and cell cycle checkpoints. Evidence indicates 14-3-3 is required for Raf1 kinase activity and phosphorylation amoung many other functions. Levels of 14-3-3 gamma in the brain are significantly increased in Alzheimer's disease and Down syndrome.

IP: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 ?g/ml. Predicted molecular weight: 30 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: 14-3-3 γ
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR