Anti-4E-BP1 Rabbit Monoclonal Antibody

Supplier: Genetex
GTX61987
89364-258EA 585.93 CAD
89364-258
Anti-4E-BP1 Rabbit Monoclonal Antibody
4E-BP1 (eIF4E-binding protein) also known as PHAS, is a 10-12 kDa acidic protein that compete with eIF4G for binding of eIF4E to the mRNA 5 cap structure . Binding of the 4E-BPs to eIF4E is reversible and is dependent on the phosphorylation status of 4E-BP1. Non-phosphorylated 4E-BP1 will bind strongly to eIF4E while, the phosphorylated form will no . Akt, TOR, MAP kinase, S6 kinase, and Cdc2 are known kinases capable of inactivating 4E-BP1 binding to eIF4E by phosphorylating either threonines 35, 45, 70 or serine 64. Although, not all phosphorylation events equally block the 4EBP1-eIF4E interaction . It has been reported previously that phosphorylation of 4E-BP1 on Thr 37 and Thr 46 is relatively insensitive to serum deprivation and rapamycin treatment, and that phosphorylation of these residues is required for the subsequent phosphorylation of a set of unidentified serum-responsive sites

Type: Primary
Antigen: 4E-BP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR