Anti-5HT5A Rabbit Polyclonal Antibody

Supplier: Genetex

GTX77999
89366-468EA 699.51 CAD
89366-468
Anti-5HT5A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to 5-HT5A Receptor

Type: Primary
Antigen: 5HT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR