Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]

Supplier: GeneTex

GTX75897
89365-932EA 689.97 CAD
89365-932
Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]
Antibodies
Mouse monoclonal antibody [33D3] to 5-Methylcytosine / 5-mC

Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: 33D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Broad
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR