Anti-6xHis Chicken Polyclonal Antibody

Supplier: Genetex
GTX77351
89353-656EA 908.9 CAD
89353-656
Anti-6xHis Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 6xHis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR