Anti-Neurofilament Light (NF-L) Mouse Monoclonal Antibody

Supplier: GeneTex
GTX27255
89362-524EA 565.34 CAD
89362-524
Anti-Neurofilament Light (NF-L) Mouse Monoclonal Antibody
Antibodies
Neurofilaments are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-L is the neurofilament light or low molecular weight polypeptide and runs on SDS-PAGE gels at about 68kDa. Antibodies to NF-L are useful for identifying neuronal cells and their processes in tissue sections and in tissue culture. NF-L antibody can also be useful in the diagnostics of neurofilament accumulations seen in many neurological diseases, such as Lou Gehrig's disease or Alzheimer's disease.

Type: Primary
Antigen: Neurofilament Light (NF-L)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bird
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR