Anti-LRP Mouse Monoclonal Antibody [clone: 5A6]

Supplier: GeneTex
GTX79838
89366-652EA 920.95 CAD
89366-652
Anti-LRP Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Mouse Monoclonal antibody to FITC labeled 85 kDa LRP [5A6] (FITC)

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: LRP
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR