Anti-LRP Mouse Monoclonal Antibody [clone: 8B8]

Supplier: GeneTex

GTX79835
89366-650EA 665.59 CAD
89366-650
Anti-LRP Mouse Monoclonal Antibody [clone: 8B8]
Antibodies
Mouse Monoclonal antibody to FITC labeled 85 kDa LRP [8B8] (FITC)

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: LRP
Clonality: Monoclonal
Clone: 8B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR