Anti-ABCG8 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX30466
89352-660EA 911.09 CAD
89352-660
Anti-ABCG8 Rabbit Polyclonal Antibody
Antibodies
Polyclonal anti - ABCG8 ( ATP Binding cassette transporter )

Not tested in other applictions. Dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ABCG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR