Anti-ACHE Goat Polyclonal Antibody

Supplier: GeneTex

GTX27396
89362-578EA 416.03 CAD
89362-578
Anti-ACHE Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Acetyl Cholinesterase

Recommended Starting Dilutions:WB: Use at 1:500 - 1:5,000ELISA: Use at 1:5,000 - 1:20,000IP: Use at 1:100

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR