Anti-ACT1 Mouse Monoclonal Antibody

GTX17853
89351-412EA 523.35 CAD
89351-412
Anti-ACT1 Mouse Monoclonal Antibody
This antibody reacts with alpha-smooth muscle as well as alpha-skeletal and alpha-cardiac (sarcomeric) isoform of actin. It is a pan muscle actin and reacts with tumors arising from smooth muscle (leiomyosarcomas) as well as skeletal muscle (rhabdomyosarcomas).

Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR