Anti-ACVR2 Mouse Monoclonal Antibody [clone: 149/1]

Supplier: GeneTex

GTX18103
89360-400EA 640.7 CAD
89360-400
Anti-ACVR2 Mouse Monoclonal Antibody [clone: 149/1]
Antibodies
ACTR2 is a serine/threonine kinase receptor. Activin signal transduction pathways are implicated in diverse developmental and physiological processes.

IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 57 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ACVR2
Clonality: Monoclonal
Clone: 149/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR