Anti-ADA Mouse Monoclonal Antibody [clone: ADA-1]

Supplier: GeneTex

GTX18104
89360-402EA 624.19 CAD
89360-402
Anti-ADA Mouse Monoclonal Antibody [clone: ADA-1]
Antibodies
This gene encodes an enzyme that catalyzes the hydrolysis of adenosine to inosine. Various mutations have been described for this gene and have been linked to human diseases. Deficiency in this enzyme causes a form of severe combined immunodeficiency disease (SCID), in which there is dysfunction of both B and T lymphocytes with impaired cellular immunity and decreased production of immunoglobulins, whereas elevated levels of this enzyme have been associated with congenital hemolytic anemia. [provided by RefSeq]

IP: Use at an assay dependent dilution. RIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 40 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ADA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR