Anti-ADA Rabbit Monoclonal Antibody

GTX62770
89364-592EA 556.26 CAD
89364-592
Anti-ADA Rabbit Monoclonal Antibody
ADA is an enzyme that catalyzes the hydrolysis of adenosine to inosine. Various mutations have been described for this protein and have been linked to human diseases. Deficiency in this enzyme causes a form of severe combined immunodeficiency disease (SCID), in which there is dysfunction of both B and T lymphocytes with impaired cellular immunity and decreased production of immunoglobulins, whereas elevated levels of this enzyme have been associated with congenital hemolytic anemia.

Type: Primary
Antigen: ADA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR