Anti-ADCY2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX14777
89367-396EA 780.52 CAD
89367-396
Anti-ADCY2 Rabbit Polyclonal Antibody
Antibodies
ADCY2 is a membrane-bound, calmodulin-insensitive adenylyl cyclase.

Type: Primary
Antigen: ADCY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR