Anti-ADA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX40578
89363-246EA 1369.12 CAD
89363-246
Anti-ADA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Adenosine Deaminase (HRP)

  • ELISA: 1:1,0000 - 1:100,000. FACS: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 104 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR