Anti-HADV Mouse Monoclonal Antibody [clone: 8C4]

Supplier: GeneTex
GTX28249
89352-360EA 1147.57 CAD
89352-360
Anti-HADV Mouse Monoclonal Antibody [clone: 8C4]
Antibodies
This antibody is used for the detection of human adenovirus. This antibody detects a different epitope than the monoclonal, GT28251.

Type: Primary
Antigen: AdenoVirus Hexon
Clonality: Monoclonal
Clone: [8C4]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR