Anti-AdV 5 E1A Mouse Monoclonal Antibody

GTX17803
89351-390EA 557 CAD
89351-390
Anti-AdV 5 E1A Mouse Monoclonal Antibody
The early region (E1A) of the adenovirus genome plays a central role in cellular transformation and regulation of gene expression. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.

Type: Primary
Antigen: AdV 5 E1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR