Anti-AGRN Mouse Monoclonal Antibody [clone: Agr-247]

Supplier: GeneTex
GTX12364
89359-350EA 754.53 CAD
89359-350
Anti-AGRN Mouse Monoclonal Antibody [clone: Agr-247]
Antibodies
Agrin is an essential extracellular matrix component which promotes clustering of nicotinic acetylcholine receptors (nAChRs) and other proteins during development at the neuromuscular junction. Agrin, MuSK and Rapsyn are all essential components for AChR aggregation, through an unknown mechanism. The C-terminal region of agrin is released into the medium, interacts with receptors on the muscle surface and induces AChR aggregation. The central region contains two O-linked glycosylation sites and a domain homologous to domain III of laminin. The N-terminal region anchors agrin to the extracellular matrix via other basal membrane components. This region also contains a protease inhibitor domain and glycosaminoglycan attachment sites; increasing the predicted MW from 200kDa to ~600kDa.

ICC: Use at a concentration of 2 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: AGRN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR